siRNA Details
| Gene Symbol: Slc12a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079690.1 | NP_001073158.1 | 25 | 3210 | ||
| NM_183354.2 | NP_899197.2 | 25 | 3210 | |||
| GenBank mRNA | BC016888.1 | - | 24 | 3188 | 4236020 | |
| BC051100.1 | - | 18 | 1974 | |||
| U20973.1 | - | 25 | 3208 | |||
| U20974.1 | - | 25 | 3208 | |||
| U20975.1 | - | 25 | 3208 | |||
| U94518.1 | - | 24 | 3157 | |||

Slc12a1
Genomic Map