siRNA Details
| Gene Symbol: Slc12a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009194.3 | NP_033220.2 | 27 | 3676 | ||
| GenBank mRNA | AK134606.1 | - | 11 | 1104 | ||
| AK164756.1 | - | 5 | 470 | |||
| CN711123.1 | - | 6 | 553 | |||
| U13174.1 | - | 27 | 3673 | |||

Slc12a2
Genomic Map