siRNA Details
| Gene Symbol: Slc12a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011390.2 | NP_035520.1 | 22 | 2967 | ||
| GenBank mRNA | AF087436.1 | - | 22 | 2969 | ||
| AK131129.1 | - | 22 | 2921 | |||
| AK143535.1 | - | 22 | 2961 | |||
| AK149750.1 | - | 22 | 2962 | |||
| AK166215.1 | - | 22 | 2961 | |||
| AK169950.1 | - | 22 | 2957 | |||
| BC059242.1 | - | 22 | 2959 | |||
| BC060133.1 | - | 22 | 2959 | |||
| BC079677.1 | - | 18 | 4824 | |||
| BC141107.1 | - | 22 | 2927 | 9055719 | ||

Slc12a7
Genomic Map