siRNA Details
| Gene Symbol: Slc16a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011391.1 | NP_035521.1 | 6 | 1373 | ||
| GenBank mRNA | AF058054.1 | - | 6 | 1373 | ||
| AK076612.1 | - | 5 | 1195 | |||
| BC089606.1 | - | 5 | 1143 | |||
| BC117803.1 | - | 5 | 1137 | 40092297 | ||
| BC117804.1 | - | 5 | 1137 | 40092299 | ||
| BC126960.1 | - | 5 | 1137 | 40092496 | ||
| CT010191.1 | - | 3 | 936 | |||

Slc16a7
Genomic Map