siRNA Details
| Gene Symbol: Slc17a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170638.1 | NP_001164109.1 | 9 | 1055 | ||
| NM_009198.3 | NP_033224.3 | 9 | 1147 | |||
| GenBank mRNA | AI663290.1 | - | 5 | 303 | ||
| AK164712.1 | - | 9 | 1057 | |||
| BC013445.1 | - | 9 | 1088 | 4241557 | ||
| X77241.1 | - | 9 | 1014 | |||

Slc17a1
Genomic Map