siRNA Details
| Gene Symbol: Slc18a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021712.2 | NP_068358.2 | 1 | 1325 | ||
| GenBank mRNA | AK158592.1 | - | 1 | 1327 | ||
| BC120498.1 | - | 1 | 942 | 40129421 | ||
| BC137705.1 | - | 1 | 941 | 8860724 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc18a3| Gene Symbol: Slc18a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021712.2 | NP_068358.2 | 1 | 1325 | ||
| GenBank mRNA | AK158592.1 | - | 1 | 1327 | ||
| BC120498.1 | - | 1 | 942 | 40129421 | ||
| BC137705.1 | - | 1 | 941 | 8860724 | ||