siRNA Details
| Gene Symbol: TNFRSF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001561.5 | NP_001552.2 | 5 | 581 | ||
| XM_006710618.3 | XP_006710681.1 | 4 | 1228 | |||
| XM_011541386.2 | XP_011539688.1 | 4 | 596 | |||
| GenBank mRNA | BC006196.2 | - | 4 | 441 | 2924109 | |
| BP381234.1 | - | 5 | 581 | |||
| L12964.2 | - | 4 | 459 | |||
| U03397.1 | - | 4 | 439 | |||

TNFRSF9
Genomic Map