siRNA Details
| Gene Symbol: Slc20a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011394.3 | NP_035524.2 | 4 | 996 | ||
| GenBank mRNA | AF196476.1 | - | 3 | 764 | ||
| AK076380.1 | - | 1 | 1656 | |||
| AK087507.1 | - | 4 | 821 | |||
| AK157206.1 | - | 4 | 870 | |||
| AK170985.1 | - | 4 | 962 | |||
| AK220047.1 | - | 1 | 25 | |||
| BC046510.1 | - | 4 | 919 | 4035182 | ||
| BC094562.1 | - | 4 | 1376 | |||
| U62559.1 | - | 3 | 475 | |||

Slc20a2
Genomic Map