siRNA Details
| Gene Symbol: Slc25a17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011399.3 | NP_035529.1 | 5 | 459 | ||
| GenBank mRNA | AJ006341.1 | - | 5 | 471 | ||
| AK002388.1 | - | 5 | 460 | |||
| AK003672.1 | - | 5 | 446 | |||
| BC008571.1 | - | 5 | 461 | 3587806 | ||
| BC011292.1 | - | 5 | 441 | 3962111 | ||

Slc25a17
Genomic Map