siRNA Details
| Gene Symbol: Slc2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011400.3 | NP_035530.2 | 6 | 1064 | ||
| GenBank mRNA | AK089353.1 | - | 6 | 1006 | ||
| AK155252.1 | - | 6 | 1006 | |||
| AK170425.1 | - | 6 | 1006 | |||
| AK170873.1 | - | 6 | 1006 | |||
| BC055340.1 | - | 6 | 1033 | 6808315 | ||
| M22998.1 | - | 6 | 968 | |||
| M23384.1 | - | 6 | 1043 | |||

Slc2a1
Genomic Map