siRNA Details
| Gene Symbol: Slc2a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009204.2 | NP_033230.2 | 10, 11 | 1542 | ||
| GenBank mRNA | AB008453.1 | - | 10, 11 | 1340 | ||
| AK137607.1 | - | 10, 11 | 1527 | |||
| AK160882.1 | - | 8, 9 | 2167 | |||
| AK164310.1 | - | 10, 11 | 1517 | |||
| BC014282.1 | - | 10, 11 | 1512 | 4207674 | ||

Slc2a4
Genomic Map