siRNA Details
| Gene Symbol: Slc34a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011402.3 | NP_035532.2 | 3 | 280 | ||
| GenBank mRNA | AK004832.1 | - | 3 | 279 | ||
| AK145599.1 | - | 3 | 279 | |||
| AK164786.1 | - | 3 | 281 | |||
| BB873139.1 | - | 3 | 281 | |||
| BC096369.1 | - | 2 | 251 | 3962784 | ||

Slc34a2
Genomic Map