siRNA Details
Gene Symbol: Slc4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011403.2 | NP_035533.1 | 13 | 1800 | ||
GenBank mRNA | AK137542.1 | - | 13 | 1696 | ||
AK157181.1 | - | 13 | 1769 | |||
AK157833.1 | - | 13 | 1658 | |||
AK157861.1 | - | 13 | 1670 | |||
AK158143.1 | - | 13 | 1801 | |||
AK158184.1 | - | 13 | 1766 | |||
AK158243.1 | - | 13 | 1670 | |||
AK158453.1 | - | 13 | 1645 | |||
AK162616.1 | - | 13 | 1645 | |||
AK165195.1 | - | 13 | 1657 | |||
AK165267.1 | - | 13 | 1664 | |||
AK171662.1 | - | 1696 | ||||
AK171808.1 | - | 13 | 1634 | |||
AK171845.1 | - | 1696 | ||||
AK172103.1 | - | 13 | 1657 | |||
AK172316.1 | - | 13 | 1645 | |||
AK172395.1 | - | 13 | 1696 | |||
AK172456.1 | - | 13 | 1680 | |||
AK172569.1 | - | 13 | 1645 | |||
AY296129.1 | - | 13 | 1633 | |||
BC052419.1 | - | 13 | 1640 | 5702599 | ||
BC053429.1 | - | 13 | 1631 | 6506270 | ||
M29379.1 | - | 13 | 1627 | |||
X02677.1 | - | 13 | 1633 | |||
X03917.1 | - | 11 | 1477 |
