siRNA Details
| Gene Symbol: Slc4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253892.1 | NP_001240821.1 | 7 | 1104 | ||
| NM_009207.3 | NP_033233.2 | 8 | 1300 | |||
| GenBank mRNA | AK145930.1 | - | 8 | 1225 | ||
| AK147858.1 | - | 8 | 1256 | |||
| AK213497.1 | - | 3 | 314 | |||
| BC054102.1 | - | 7 | 1109 | 30017310 | ||
| J04036.1 | - | 8 | 1215 | |||

Slc4a2
Genomic Map