siRNA Details
| Gene Symbol: Slc4a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009208.3 | NP_033234.2 | 12 | 1916 | ||
| GenBank mRNA | AK147218.1 | - | 2253 | |||
| BC066086.1 | - | 13 | 1790 | |||
| BC080271.1 | - | 12 | 1911 | 6831942 | ||
| M28383.1 | - | 12 | 1886 | |||

Slc4a3
Genomic Map