siRNA Details
| Gene Symbol: Slc5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019810.4 | NP_062784.3 | 8 | 971 | ||
| GenBank mRNA | AF163846.1 | - | 8 | 733 | ||
| AF208031.1 | - | 8 | 971 | |||
| AK033512.1 | - | 8 | 957 | |||
| AK085602.1 | - | 8 | 762 | |||
| BC003845.1 | - | 8 | 951 | 3497611 | ||
| U27269.1 | - | 6 | 541 | |||

Slc5a1
Genomic Map