siRNA Details
| Gene Symbol: Slc8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001112798.2 | NP_001106269.1 | 3 | 1929 | ||
| NM_001286684.1 | NP_001273613.1 | 3 | 1929 | |||
| NM_011406.3 | NP_035536.2 | 3 | 1929 | |||
| GenBank mRNA | AK048160.1 | - | 1496 | |||
| BC079673.1 | - | 2 | 1820 | 30532156 | ||
| DQ388998.1 | - | 2 | 1819 | |||
| U70033.1 | - | 3 | 1919 | |||

Slc8a1
Genomic Map