siRNA Details
| Gene Symbol: Slfn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011407.1 | NP_035537.1 | 2 | 463 | ||
| GenBank mRNA | AF099972.1 | - | 2 | 463 | ||
| AK138710.1 | - | 2 | 466 | |||
| BC052869.1 | - | 2 | 471 | 30073520 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slfn1| Gene Symbol: Slfn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011407.1 | NP_035537.1 | 2 | 463 | ||
| GenBank mRNA | AF099972.1 | - | 2 | 463 | ||
| AK138710.1 | - | 2 | 466 | |||
| BC052869.1 | - | 2 | 471 | 30073520 | ||