siRNA Details
| Gene Symbol: Slfn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011408.1 | NP_035538.1 | 2 | 285 | ||
| GenBank mRNA | AF099973.1 | - | 2 | 285 | ||
| BC138749.1 | - | 2 | 240 | 8861771 | ||
| BC138750.1 | - | 2 | 240 | 8861772 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slfn2| Gene Symbol: Slfn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011408.1 | NP_035538.1 | 2 | 285 | ||
| GenBank mRNA | AF099973.1 | - | 2 | 285 | ||
| BC138749.1 | - | 2 | 240 | 8861771 | ||
| BC138750.1 | - | 2 | 240 | 8861772 | ||