siRNA Details
| Gene Symbol: Slit3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011412.3 | NP_035542.2 | 15, 16 | 1668 | ||
| GenBank mRNA | AF144629.1 | - | 15, 16 | 1609 | ||
| AK147292.1 | - | 15, 16 | 1669 | |||
| BC150780.1 | - | 15, 16 | 1645 | 9087691 | ||

Slit3
Genomic Map