siRNA Details
| Gene Symbol: Hltf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009210.3 | NP_033236.2 | 17 | 1995 | ||
| GenBank mRNA | AB221636.1 | - | 17 | 1818 | ||
| AF010600.1 | - | 17 | 1995 | |||
| AF165911.1 | - | 17 | 1974 | |||
| AK141235.1 | - | 17 | 1988 | |||
| BC057116.1 | - | 17 | 1964 | 6408672 | ||
| BC059240.1 | - | 17 | 1964 | 6408672 | ||

Hltf
Genomic Map