siRNA Details
| Gene Symbol: Kdm5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013668.4 | NP_038696.2 | 9 | 1501 | ||
| GenBank mRNA | AF127245.1 | - | 9 | 1456 | ||
| AK129096.1 | - | 6 | 675 | |||
| AK150152.1 | - | 9 | 1502 | |||
| AK155279.1 | - | 9 | 1486 | |||
| AK155427.1 | - | 9 | 1486 | |||
| AK155651.1 | - | 9 | 1499 | |||
| AK158340.1 | - | 9 | 1486 | |||
| BC043096.1 | - | 8 | 1333 | 6405943 | ||
| BC054550.1 | - | 8 | 1333 | 6405943 | ||
| Z29651.1 | - | 9 | 1063 | |||

Kdm5c
Genomic Map