siRNA Details
| Gene Symbol: Sstr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009219.3 | NP_033245.2 | 1 | 472 | ||
| GenBank mRNA | AK051189.1 | - | 507 | |||
| BC138487.1 | - | 451 | 8861507 | |||
| BC138488.1 | - | 451 | 8861508 | |||
More Information
Set Membership: |
|

Sstr4
Genomic Map