siRNA Details
| Gene Symbol: Siglec1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011426.3 | NP_035556.3 | 12 | 3706 | ||
| GenBank mRNA | BC141335.1 | - | 12 | 3300 | 9055947 | |
| BC141336.1 | - | 12 | 3296 | 9055948 | ||
| Z36233.1 | - | 12 | 3701 | |||
| Z36293.1 | - | 12 | 3701 | |||

Siglec1
Genomic Map