siRNA Details
| Gene Symbol: Snn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009223.3 | NP_033249.1 | 2 | 421 | ||
| GenBank mRNA | AK149685.1 | - | 2 | 370 | ||
| BC006961.1 | - | 2 | 389 | 3707801 | ||
| BC017685.1 | - | 2 | 338 | 4241981 | ||
| BC038475.1 | - | 1 | 462 | 3157762 | ||
| BC063027.1 | - | 2 | 358 | 5343626 | ||
| BC087932.1 | - | 366 | 6706156 | |||
| CJ166486.1 | - | 2 | 373 | |||

Snn
Genomic Map