siRNA Details
| Gene Symbol: Snrpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009225.2 | NP_033251.1 | 3 | 386 | ||
| GenBank mRNA | AK077537.1 | - | 3 | 339 | ||
| AK146319.1 | - | 3 | 429 | |||
| AK178243.1 | - | 2 | 186 | |||
| AK186005.1 | - | 2 | 186 | |||
| AK203845.1 | - | 2 | 186 | |||
| AK207139.1 | - | 2 | 186 | |||
| BG086527.2 | - | 3 | 344 | |||

Snrpb
Genomic Map