siRNA Details
| Gene Symbol: Snrpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009226.4 | NP_033252.1 | 3 | 358 | ||
| GenBank mRNA | AK005517.1 | - | 3 | 348 | ||
| AK007674.1 | - | 3 | 359 | |||
| AK011455.1 | - | 3 | 352 | |||
| AK013075.1 | - | 3 | 325 | |||
| AK134582.1 | - | 3 | 352 | |||
| AK135979.1 | - | 3 | 348 | |||
| BC024875.1 | - | 3 | 344 | 4009014 | ||
| M58558.1 | - | 3 | 287 | |||

Snrpd1
Genomic Map