siRNA Details
| Gene Symbol: Snta1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009228.2 | NP_033254.2 | 3 | 609 | ||
| GenBank mRNA | AK137379.1 | - | 3 | 610 | ||
| BB625367.1 | - | 3 | 602 | |||
| BC018546.1 | - | 3 | 559 | 3674921 | ||
| U00677.1 | - | 3 | 606 | |||

Snta1
Genomic Map