siRNA Details
| Gene Symbol: Son | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019973.2 | NP_064357.2 | 3 | 3346 | ||
| NM_178880.4 | NP_849211.3 | 3 | 3346 | |||
| GenBank mRNA | AB546195.1 | - | 4 | 3161 | ||
| AF193607.1 | - | 4 | 3170 | |||
| BC046419.1 | - | 3 | 3285 | |||
| BC145121.1 | - | 3 | 3288 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001170327, NM_001170328, XM_006248064, XM_006248065, XM_008768580.

Son
Genomic Map