siRNA Details
| Gene Symbol: Son | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019973.2 | NP_064357.2 | 3 | 5201 | ||
| NM_178880.4 | NP_849211.3 | 3 | 5201 | |||
| GenBank mRNA | AB546195.1 | - | 4 | 5016 | ||
| AF193607.1 | - | 4 | 5025 | |||
| AK034916.1 | - | 1 | 1363 | |||
| AK040621.1 | - | 1 | 204 | |||
| BC003245.1 | - | 1 | 1579 | |||
| BC046419.1 | - | 3 | 5140 | |||
| BC145121.1 | - | 3 | 5143 | |||

Son
Genomic Map