siRNA Details
| Gene Symbol: Son | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019973.2 | NP_064357.2 | 3 | 5447 | ||
| NM_178880.4 | NP_849211.3 | 3 | 5447 | |||
| GenBank mRNA | AB546195.1 | - | 4 | 5262 | ||
| AF193607.1 | - | 4 | 5271 | |||
| AK034916.1 | - | 1 | 1609 | |||
| AK040621.1 | - | 1 | 450 | |||
| BC003245.1 | - | 1 | 1825 | |||
| BC046419.1 | - | 3 | 5386 | |||
| BC145121.1 | - | 3 | 5389 | |||

Son
Genomic Map