siRNA Details
| Gene Symbol: Sp100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013673.3 | NP_038701.2 | 12 | 1069 | ||
| GenBank mRNA | AK083304.1 | - | 12 | 1070 | ||
| AK152348.1 | - | 11 | 1052 | |||
| BC069183.1 | - | 11 | 1038 | 4911025 | ||
| CT010199.1 | - | 12 | 1017 | |||
| U83630.1 | - | 12 | 1106 | |||
| U83636.1 | - | 11 | 963 | |||
| U83637.1 | - | 2 | 89 | |||

Sp100
Genomic Map