siRNA Details
| Gene Symbol: INCENP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040694.1 | NP_001035784.1 | 18 | 2608 | ||
| NM_020238.2 | NP_064623.2 | 17 | 2596 | |||
| XM_006718533.2 | XP_006718596.1 | 18 | 2582 | |||
| XM_011544995.2 | XP_011543297.1 | 18 | 2615 | |||
| XM_011544996.2 | XP_011543298.1 | 18 | 2603 | |||
| XM_011544997.2 | XP_011543299.1 | 17 | 2563 | |||
| XM_011544998.2 | XP_011543300.1 | 17 | 2591 | |||
| GenBank mRNA | AY714053.1 | - | 17 | 2406 | ||
| BC098576.1 | - | 18 | 2608 | 30924748 | ||

INCENP
Genomic Map