siRNA Details
| Gene Symbol: Serpinb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009256.3 | NP_033282.1 | 4, 5 | 584 | ||
| GenBank mRNA | AK035624.1 | - | 4, 5 | 523 | ||
| AK087785.1 | - | 4, 5 | 493 | |||
| AK161581.1 | - | 4, 5 | 585 | |||
| BC029900.1 | - | 4, 5 | 512 | 4925100 | ||
| U96700.1 | - | 4, 5 | 463 | |||

Serpinb9
Genomic Map