siRNA Details
| Gene Symbol: Spr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011467.2 | NP_035597.2 | 1 | 130 | ||
| GenBank mRNA | AK004941.1 | - | 1 | 122 | ||
| AK154720.1 | - | 133 | ||||
| BC010400.1 | - | 1 | 127 | 4219245 | ||
| BC054423.1 | - | 1 | 114 | 4985039 | ||
| BY113096.1 | - | 1 | 131 | |||
| CK620382.1 | - | 1 | 98 | |||
| S77493.1 | - | 1 | 132 | |||

Spr
Genomic Map