siRNA Details
| Gene Symbol: Spr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011467.2 | NP_035597.2 | 3 | 757 | ||
| GenBank mRNA | AK004941.1 | - | 3 | 567 | ||
| AK207238.1 | - | 2 | 162 | |||
| AK216452.1 | - | 2 | 162 | |||
| BC010400.1 | - | 3 | 754 | 4219245 | ||
| BC054423.1 | - | 3 | 741 | 4985039 | ||
| S71375.1 | - | 1 | 116 | |||
| S77493.1 | - | 3 | 759 | |||
| W65601.1 | - | 2 | 216 | |||
| Z21947.1 | - | 1 | 116 | |||

Spr
Genomic Map