siRNA Details
Gene Symbol: Sprr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009265.3 | NP_033291.1 | 2 | 716 | ||
GenBank mRNA | AK003398.1 | - | 2 | 717 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sprr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009265.3 | NP_033291.1 | 2 | 716 | ||
GenBank mRNA | AK003398.1 | - | 2 | 717 |