siRNA Details
| Gene Symbol: Sephs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009266.3 | NP_033292.2 | 1 | 1312 | ||
| GenBank mRNA | AK077573.1 | - | 1 | 1314 | ||
| AK149516.1 | - | 1 | 1309 | |||
| AK168722.1 | - | 1 | 1313 | |||
| AK168965.1 | - | 1 | 1313 | |||
| BC016508.1 | - | 1 | 1304 | 4911695 | ||
| BC028966.1 | - | 1 | 1289 | 5374184 | ||
| BC043334.1 | - | 1 | 1293 | 5100207 | ||
| U43285.1 | - | 1 | 1309 | |||

Sephs2
Genomic Map