siRNA Details
| Gene Symbol: Spt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009267.2 | NP_033293.2 | 3 | 332 | ||
| GenBank mRNA | AK006124.1 | - | 3 | 330 | ||
| BC030164.1 | - | 3 | 325 | |||
| BC106151.1 | - | 3 | 316 | 4923478 | ||
| CK616968.1 | - | 3 | 333 | |||
| M33974.1 | - | 3 | 328 | |||
| X00349.1 | - | 3 | 329 | |||

Spt1
Genomic Map