siRNA Details
| Gene Symbol: Mucl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009268.1 | NP_033294.1 | 525 | |||
| GenBank mRNA | BC012233.1 | - | 515 | 4165141 | ||
| BC024641.1 | - | 516 | 4165428 | |||
| BC059095.1 | - | 525 | 4925159 | |||
| BC091766.1 | - | 457 | 4190032 | |||
| BC093507.1 | - | 525 | 4921535 | |||
| M33975.1 | - | 525 | ||||

Mucl1
Genomic Map