siRNA Details
| Gene Symbol: Sqle | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009270.3 | NP_033296.1 | 2, 3 | 1289 | ||
| GenBank mRNA | AK138457.1 | - | 2, 3 | 1300 | ||
| AK148419.1 | - | 2, 3 | 1305 | |||
| AK161837.1 | - | 2, 3 | 1275 | |||
| AK163502.1 | - | 2, 3 | 1269 | |||
| AK179451.1 | - | 1, 2 | 172 | |||
| AK181007.1 | - | 1, 2 | 172 | |||
| BC042781.1 | - | 2, 3 | 546 | 4915911 | ||
| BC056361.1 | - | 2, 3 | 1289 | 6816800 | ||
| D42048.1 | - | 2, 3 | 1155 | |||

Sqle
Genomic Map