siRNA Details
| Gene Symbol: Sqle | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009270.3 | NP_033296.1 | 6 | 1780 | ||
| GenBank mRNA | AK138457.1 | - | 6 | 1791 | ||
| AK148419.1 | - | 6 | 1765 | |||
| AK161837.1 | - | 6 | 1766 | |||
| AK163502.1 | - | 6 | 1760 | |||
| AK189190.1 | - | 1 | 69 | |||
| AK210458.1 | - | 1 | 37 | |||
| AK211050.1 | - | 1 | 30 | |||
| BC042781.1 | - | 6 | 1037 | 4915911 | ||
| BC056361.1 | - | 6 | 1780 | 6816800 | ||
| D42048.1 | - | 6 | 1646 | |||

Sqle
Genomic Map