siRNA Details
| Gene Symbol: Srebf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033218.1 | NP_150087.1 | 10 | 2006 | ||
| GenBank mRNA | AK155857.1 | - | 11 | 1954 | ||
| AK172219.1 | - | 10 | 2013 | |||
| BQ963055.1 | - | 1 | 61 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srebf2| Gene Symbol: Srebf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033218.1 | NP_150087.1 | 10 | 2006 | ||
| GenBank mRNA | AK155857.1 | - | 11 | 1954 | ||
| AK172219.1 | - | 10 | 2013 | |||
| BQ963055.1 | - | 1 | 61 | |||