siRNA Details
| Gene Symbol: INHBB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002193.2 | NP_002184.2 | 2 | 788 | ||
| GenBank mRNA | BC030029.2 | - | 2 | 788 | 5258851 | |
| M13437.1 | - | 2 | 582 | |||
| M31682.1 | - | 2 | 681 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

INHBB| Gene Symbol: INHBB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002193.2 | NP_002184.2 | 2 | 788 | ||
| GenBank mRNA | BC030029.2 | - | 2 | 788 | 5258851 | |
| M13437.1 | - | 2 | 582 | |||
| M31682.1 | - | 2 | 681 | |||