siRNA Details
Gene Symbol: Srm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009272.4 | NP_033298.1 | 5, 6 | 870 | ||
GenBank mRNA | AK028359.1 | - | 5, 6 | 881 | ||
AK051101.1 | - | 5, 6 | 866 | |||
AK146395.1 | - | 5, 6 | 827 | |||
BC005566.1 | - | 5, 6 | 874 | 3966710 | ||
L19311.1 | - | 5, 6 | 838 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
