siRNA Details
| Gene Symbol: Srpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016795.3 | NP_058075.2 | 11 | 1184 | ||
| GenBank mRNA | AB012290.1 | - | 10 | 1142 | ||
| AJ224115.1 | - | 11 | 1178 | |||
| AK015754.1 | - | 7 | 712 | |||
| AK140761.1 | - | 594 | ||||
| AK151152.1 | - | 11 | 1247 | |||
| AK164733.1 | - | 1 | 1169 | |||
| BC005707.1 | - | 11 | 1153 | 3157835 | ||
| BC050761.1 | - | 11 | 1184 | 6771151 | ||

Srpk1
Genomic Map