siRNA Details
| Gene Symbol: Srpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016795.3 | NP_058075.2 | 4 | 267 | ||
| GenBank mRNA | AB012290.1 | - | 3 | 225 | ||
| AJ224115.1 | - | 4 | 261 | |||
| AK017187.1 | - | 4 | 264 | |||
| AK141271.1 | - | 4 | 334 | |||
| AK151152.1 | - | 4 | 330 | |||
| AK161346.1 | - | 4 | 245 | |||
| AK161388.1 | - | 4 | 246 | |||
| AK198647.1 | - | 3 | 220 | |||
| BC005707.1 | - | 4 | 236 | 3157835 | ||
| BC050761.1 | - | 4 | 267 | 6771151 | ||

Srpk1
Genomic Map