siRNA Details
| Gene Symbol: Nhp2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011482.4 | NP_035612.2 | 2 | 186 | ||
| GenBank mRNA | AK004489.1 | - | 2 | 134 | ||
| AK134943.1 | - | 124 | ||||
| AK145391.1 | - | 2 | 188 | |||
| AK145608.1 | - | 2 | 168 | |||
| AK168808.1 | - | 2 | 156 | |||
| AK168930.1 | - | 2 | 139 | |||
| AK169103.1 | - | 2 | 170 | |||
| BC026755.1 | - | 2 | 132 | 3154066 | ||
| BC054450.1 | - | 2 | 132 | 6491298 | ||
| BC083315.1 | - | 2 | 108 | 5714819 | ||
| U95114.1 | - | 2 | 158 | |||

Nhp2l1
Genomic Map