siRNA Details
| Gene Symbol: Stat6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009284.2 | NP_033310.2 | 3 | 429 | ||
| GenBank mRNA | AF481808.1 | - | 2 | 144 | ||
| AF481809.1 | - | 2 | 144 | |||
| AK149563.1 | - | 3 | 389 | |||
| AK163053.1 | - | 3 | 430 | |||
| AK217738.1 | - | 2 | 172 | |||
| BC127054.1 | - | 3 | 219 | 40109712 | ||

Stat6
Genomic Map