siRNA Details
| Gene Symbol: Stau1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109905.2 | NP_001103375.1 | 10 | 1591 | ||
| NM_001109906.2 | NP_001103376.1 | 10 | 1609 | |||
| NM_001291160.1 | NP_001278089.1 | 10 | 1591 | |||
| NM_011490.4 | NP_035620.1 | 10 | 1585 | |||
| GenBank mRNA | AF061942.1 | - | 10 | 1585 | ||
| AF395842.1 | - | 9 | 1312 | |||
| AK005002.2 | - | 10 | 1532 | |||
| AK139532.1 | - | 10 | 1538 | |||
| AK145826.1 | - | 10 | 1576 | |||
| AK151835.1 | - | 10 | 1397 | |||
| AK152269.1 | - | 10 | 1403 | |||
| AK152744.1 | - | 10 | 1397 | |||
| AK158952.1 | - | 5 | 2172 | |||
| AK159602.1 | - | 10 | 1526 | |||
| AK159781.1 | - | 10 | 1561 | |||
| AK160979.1 | - | 10 | 1550 | |||
| AK167637.1 | - | 10 | 1532 | |||
| AY391773.1 | - | 10 | 1257 | |||
| BC012959.1 | - | 10 | 1555 | 4188394 | ||
| BC082277.1 | - | 10 | 1528 | |||

Stau1
Genomic Map